missing translation for 'onlineSavingsMsg'
Learn More

Didodecyl Phthalate, TRC

Product Code. 17988664
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
Unit Size:
100mg
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17988664 100 mg 100mg
17900075 1 g 1g
17929734 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17988664 Supplier Toronto Research Chemicals Supplier No. TRCD441765100MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2432-90-8
Molecular Formula C32 H54 O4
Molecular Weight (g/mol) 502.77
Synonym Phthalic acid, bis-dodecyl ester, 1,2-Benzenedicarboxylic acid, 1,2-didodecyl ester (ACI), 1,2-Benzenedicarboxylic acid, didodecyl ester (9CI), Phthalic acid, didodecyl ester (6CI, 7CI, 8CI), Bis-didecyl phthalate, Di-n-dodecyl phthalate, Didodecyl phthalate, Dilauryl phthalate, NSC 68022, Vinycizer 126
IUPAC Name didodecyl benzene-1,2-dicarboxylate
SMILES CCCCCCCCCCCCOC(=O)c1ccccc1C(=O)OCCCCCCCCCCCC

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C32H54O4/c1-3-5-7-9-11-13-15-17-19-23-27-35-31(33)29-25-21-22-26-30(29)32(34)36-28-24-20-18-16-14-12-10-8-6-4-2/h21-22,25-26H,3-20,23-24,27-28H2,1-2H3
Formula Weight 502.4022
Chemical Name or Material Phthalic acid, bis-dodecyl ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.