Learn More
MedChemExpress DiD perchlorate
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Specifications
Specifications
| Content And Storage | 4°C, protect from light, stored under nitrogen ∣ The compound is unstable in solutions, freshly prepared is recommended. |
| Quantity | 5 mg |
| Shipping Condition | Room temperature |
| Solubility | DMSO : 25 mg/mL (26.04 mM; ultrasonic and warming and heat to 60°C) |
| Target | HBV |
| Product Type | Dye Reagents |
| Molecular Weight (g/mol) | 959.9 g/mol |
| CAS | 127274-91-3 |