missing translation for 'onlineSavingsMsg'
Learn More

Dictamnine, TRC

Product Code. 30382336
Change view
Click to view available options
Quantity:
10 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30382336 25 mg
30380870 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30382336 Supplier Toronto Research Chemicals Supplier No. TRCD43658525MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 484-29-7
Molecular Formula C12H9NO2
Molecular Weight (g/mol) 199.21
Synonym Dictamnine, 4-Methoxyfuro[2,3-b]quinoline, Dectamine, Dictamine
SMILES COC1=C2C(OC=C2)=NC3=CC=CC=C13

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C12H9NO2/c1-14-11-8-4-2-3-5-10(8)13-12-9(11)6-7-15-12/h2-7H,1H3
Chemical Name or Material Dictamnine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.