Learn More
Dicobalt Edetate (>90%), TRC
Product Code. 30381131
Change view
Click to view available options
Quantity:
500 mg
5 g
Unit Size:
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30381131 5 g -
30376948 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30381131 Supplier Toronto Research Chemicals Supplier No. TRCD4366805G
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 36499-65-7
Molecular Formula C10H12Co2N2O8
Molecular Weight (g/mol) 406.08
Synonym Cobalt [(Ethylenedinitrilo)tetraacetato]cobaltate(II), Ba 2724, Cobalt(2)-edathamil, Dicobalt EDTA, Dicobalt Ethylenediaminetetraacetate, EDTA Cobalt Salt, Kelocyanor, (OC-6-21)-[[N,N'-1,2-Ethanediylbis[N-(carboxymethyl)glycinato]](4-)-N,N',O,O',ON,ON']cobaltate(2-) Cobalt(2+) (1:1)
IUPAC Name 2-[2-[bis(carboxylatomethyl)amino]ethyl-(carboxylatomethyl)amino]acetate;cobalt(2+)
SMILES CCCCCC/C=C\CCCCCCCC(=O)OCC1OC1
Specifications
Quantity 5 g
InChI Formula InChI=1S/C10H16N2O8.2Co/c13-7(14)3-11(4-8(15)16)1-2-12(5-9(17)18)6-10(19)20;;/h1-6H2,(H,13,14)(H,15,16)(H,17,18)(H,19,20);;/q;2*+2/p-4
Formula Weight 405.9258
Chemical Name or Material Dicobalt Edetate