Learn More
Diclosan, TRC
Product Code. 17914935
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17914935 100 mg 100mg
17904235 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17914935 Supplier Toronto Research Chemicals Supplier No. TRCD436620100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 3380-30-1
Molecular Formula C12 H8 Cl2 O2
Molecular Weight (g/mol) 255.1
Synonym Phenol, 5-chloro-2-(p-chlorophenoxy)- (7CI,8CI), 5-Chloro-2-(4-chlorophenoxy)phenol, 2-Hydroxy-4,4'-dichlorodiphenyl ether, 4,4'-Dichloro-2-hydroxydiphenyl ether, Diclosan, Soneclosan, Tinopal HP 100, Tinosan HP 100, p-Chlorophenyl 2-hydroxy-4-chlorophenyl ether
IUPAC Name 5-chloro-2-(4-chlorophenoxy)phenol
SMILES Oc1cc(Cl)ccc1Oc2ccc(Cl)cc2
Specifications
Quantity 100 mg
InChI Formula InChI=1S/C12H8Cl2O2/c13-8-1-4-10(5-2-8)16-12-6-3-9(14)7-11(12)15/h1-7,15H
Formula Weight 253.9901
Percent Purity >95%
Chemical Name or Material Diclosan
Grade HPLC