missing translation for 'onlineSavingsMsg'
Learn More

Diclofenamide, TRC

Product Code. 30134619
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134619 25 mg 25mg
30007744 5 mg 5mg
30008019 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134619 Supplier Toronto Research Chemicals Supplier No. TRCD43653025MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 120-97-8
Molecular Formula C6 H6 Cl2 N2 O4 S2
Molecular Weight (g/mol) 305.16
Synonym m-Benzenedisulfonamide, 4,5-dichloro-, 1,3-Disulfamoyl-4,5-dichlorobenzene, 1,3-Disulfamyl-4,5-dichlorobenzene, 3,4-Dichloro-5-sulfamylbenzenesulfonamide, 4,5-Dichloro-1,3-disulfamoylbenzene, 4,5-Dichloro-m-benzenedisulfonamide, Antidrasi, CB 8000, Daramide, Daranide, Dichlofenamide, Dichlorophenamide, Dichlorphenamide, Diclofenamid, Diclofenamide, Glaucol, Oratrol
IUPAC Name 4,5-dichlorobenzene-1,3-disulfonamide
SMILES NS(=O)(=O)c1cc(Cl)c(Cl)c(c1)S(=O)(=O)N

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C6H6Cl2N2O4S2/c7-4-1-3(15(9,11)12)2-5(6(4)8)16(10,13)14/h1-2H,(H2,9,11,12)(H2,10,13,14)
Formula Weight 303.9146
Chemical Name or Material Dichlorphenamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.