missing translation for 'onlineSavingsMsg'
Learn More

Dichlorphenamide, MedChemExpress

Product Code. 16438286
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16438286 100 mg 100mg
16434357 50 mg 50mg
16428478 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16438286 Supplier Medchem Express Supplier No. HYB0397
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress Dichlorphenamide (Diclofenamide) is an orally active, specific, carbonic anhydrase inhibitor. Dichlorphenamide can reduce intraocular pressure by inhibiting the secretion of water from the eye. Dichlorphenamide can be used for glaucoma research.

Dichlorphenamide (Diclofenamide) is an orally active, specific, carbonic anhydrase inhibitor. Dichlorphenamide can reduce intraocular pressure by inhibiting the secretion of water from the eye. Dichlorphenamide can be used for glaucoma research.

Specifications

Chemical Name or Material Dichlorphenamide
CAS 120-97-8
Color White
Purity Grade Notes Research
Molecular Formula C6H6Cl2N2O4S2
Quantity 100 mg
Synonym Diclofenamide
Solubility Information DMSO : 100 mg/mL (327.70 mM; Need ultrasonic) ∣H2O : 1 mg/mL (3.28 mM; ultrasonic and warming and heat to 80°C)
SMILES O=S(C1=CC(Cl)=C(Cl)C(S(=O)(N)=O)=C1)(N)=O
Molecular Weight (g/mol) 305.16
Formula Weight 305.16
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.