Learn More
Dichloromethyl Methyl Ether, Sigma-Aldrich™
Product Code. 15889010
Change view
Click to view available options
Quantity:
25 mL
Unit Size:
25mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15889010 25 mL 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15889010 Supplier Sigma Aldrich Supplier No. 8034670025
 Restricted Item
Estimated Shipment: 17-11-2026
For synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1090285
Specifications
Density 1.29 g/cm3 at 20°C
Boiling Point 85°C to 87°C/1013 hPa
MDL Number MFCD00000828
Quantity 25 mL
Synonym Dichloromethyl methyl ether, 1,1-Dichloromethylether, Dichloromethyl methyl ether
Molecular Weight (g/mol) 114.96
Grade Research