Learn More
Dichloromethane, BAKER ANALYZED™ ACS, J.T. Baker™
Product Code. 10046100 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
1 L
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10046100 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10046100 Supplier JT Baker Supplier No. 7053.1000
Exceeds ACS Specifications
Chemical Identifiers
CAS 75-09-2
Molecular Formula CH2Cl2
Molecular Weight (g/mol) 84.93
MDL Number MFCD00000881
InChI Key YMWUJEATGCHHMB-UHFFFAOYSA-N
Synonym Methylene chloride
IUPAC Name dichloromethane
SMILES ClCCl
Specifications
Melting Point -95°C
Density 1.322 g/cm³ (20°C)
Quantity 1 L
Packaging Glass Bottle
Boiling Point 39.8°C (1013 hPa)
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Carcinogenicity Category 2
Hazard Statement
  • H351-Suspected of causing cancer.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P281-Use personal protective equipment as required.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P405-Store locked up.