Learn More
Dichloroacetaldehyde Hydrate, TRC
Product Code. 30009258
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30009258 1 g 1g
30004197 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30009258 Supplier Toronto Research Chemicals Supplier No. TRCD4317651G
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 16086-14-9
Molecular Formula C2H2Cl2O.H2O
Molecular Weight (g/mol) 130.958
Synonym 2,2-Dichloro-1,1-ethanediol (ACI), Acetaldehyde, dichloro-, hydrate (6CI), Dichloroacetaldehyde hydrate
IUPAC Name 2,2-dichloroacetaldehyde;hydrate
SMILES O.ClC(Cl)C=O
Specifications
Quantity 1 g
InChI Formula InChI=1S/C2H2Cl2O.H2O/c3-2(4)1-5;/h1-2H;1H2
Formula Weight 129.9588
Percent Purity >95%
Chemical Name or Material Dichloroacetaldehyde Hydrate
Grade HPLC