missing translation for 'onlineSavingsMsg'
Learn More

Dichloroacetaldehyde Diethyl Acetal, TRC

Product Code. 30137022
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137022 10 g 10g
30136228 5 g 5g
30006244 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137022 Supplier Toronto Research Chemicals Supplier No. TRCD43172510G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 619-33-0
Molecular Formula C6 H12 Cl2 O2
Molecular Weight (g/mol) 187.064
Synonym 1,1-Dichloro-2,2-diethoxy-ethane, Dichloro-acetaldehyde Diethyl Acetal, 1,1-Dichloro-2,2-diethoxyethane, 2,2-Dichloro-1,1-diethoxyethane, Dichloroacetaldehyde Diethyl Acetal
IUPAC Name 1,1-dichloro-2,2-diethoxyethane
SMILES CCOC(OCC)C(Cl)Cl

Specifications

Quantity 10 g
InChI Formula InChI=1S/C6H12Cl2O2/c1-3-9-6(5(7)8)10-4-2/h5-6H,3-4H2,1-2H3
Formula Weight 186.021
Chemical Name or Material Dichloroacetaldehyde Diethyl Acetal
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.