missing translation for 'onlineSavingsMsg'
Learn More

Dibutyltin oxide, Sigma-Aldrich™

Product Code. 32237696
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
25 kg
missing translation for 'unitSize':
25kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity missing translation for 'unitSize'
32237696 25 kg 25kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
Product Code. 32237696 missing translation for 'mfr' Sigma Aldrich missing translation for 'supplierNo' 8035399025
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

For synthesis

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dibutyltin oxide
CAS 818-08-6
Density 1.5 g/cm³ at 20°C
Boiling Point >300°C (decomposes)
Molecular Formula (CH3CH2CH2CH2)2SnO
Quantity 25 kg
Synonym Dibutyltin oxide, Dibutyloxostannane, Dibutyltin oxide
Molecular Weight (g/mol) 248.94
Grade Not Available
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.