Learn More
Dibutyltin Diacetate, Sigma-Aldrich™
Product Code. 15894521
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15894521 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15894521 Supplier Sigma Aldrich Supplier No. 8203860100
 Restricted Item
Estimated Shipment: 12-11-2026
For synthesis
TRUSTED_SUSTAINABILITY
Specifications
CAS 1067-33-0
Melting Point 12°C
Density 1.30 g/cm3 at 20°C
Boiling Point 142°C to 145°C/13 hPa
Molecular Formula (CH3CH2CH2CH2)2Sn(OOCCH3)2
MDL Number MFCD00008697
Quantity 100 mL
Synonym Dibutyltin diacetate, Diacetoxydibutylstannane, Diacetoxydibutyltin, Dibutyltin diacetate
Molecular Weight (g/mol) 351.03
Grade Research
Chemical Name or Material Dibutyltin Diacetate