Learn More
Dibutyl succinate, 99%
Product Code. 15230213 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
100 mL
Unit Size:
100mL
25mL
5mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15230213 25 mL 25mL
15289826 5 mL 5mL
15299826 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15230213 Supplier Thermo Scientific Acros Supplier No. 453010250
Request Bulk or Custom Format
CAS: 141-03-7 | C12H22O4 | 230.3 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 141-03-7
Molecular Formula C12H22O4
Molecular Weight (g/mol) 230.3
InChI Key YUXIBTJKHLUKBD-UHFFFAOYSA-N
Synonym dibutyl succinate, tabutrex, succinic acid di-n-butyl ester, di-n-butyl succinate, tabatrex, dibutylsuccinate, butyl butanedioate, butanedioic acid, dibutyl ester, succinic acid dibutyl ester, dnbs
PubChem CID 8830
ChEBI CHEBI:82263
IUPAC Name dibutyl butanedioate
SMILES CCCCOC(=O)CCC(=O)OCCCC
Specifications
Melting Point -29.0°C
Density 0.9800g/mL
Boiling Point 274.0°C
Flash Point 144°C
Assay Percent Range 98.5% min. (GC)
Linear Formula C4H9O2CH2CH2CO2C4H9
Quantity 25 mL
Specific Gravity 0.98
Solubility Information Solubility in water: slightly.
Formula Weight 230.3
Percent Purity 99%
Chemical Name or Material Dibutyl succinate