missing translation for 'onlineSavingsMsg'
Learn More

Dibromoiodomethane (90%), TRC

Product Code. 17982595
Change view
Click to view available options
Quantity:
25 mg
250 mg
Unit Size:
250mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17982595 25 mg 25mg
17961705 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17982595 Supplier Toronto Research Chemicals Supplier No. TRCD42550025MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 593-94-2
Molecular Formula C H Br2 I
Molecular Weight (g/mol) 299.73
Synonym Methane, dibromoiodo- (6CI,7CI,9CI), Dibromoiodomethane
IUPAC Name dibromo(iodo)methane
SMILES BrC(Br)I

Specifications

Quantity 25 mg
InChI Formula InChI=1S/CHBr2I/c2-1(3)4/h1H
Formula Weight 297.749
Chemical Name or Material Dibromoiodomethane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.