missing translation for 'onlineSavingsMsg'
Learn More

Dibromoacetamide, TRC

Product Code. 30040660
Change view
Click to view available options
Quantity:
25 mg
250 mg
1 g
Unit Size:
1g
250mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30040660 25 mg 25mg
30040942 1 g 1g
30040657 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30040660 Supplier Toronto Research Chemicals Supplier No. TRCD42357025MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 598-70-9
Molecular Formula C2 H3 Br2 N O
Molecular Weight (g/mol) 216.86
Synonym 2,2-Dibromoacetamide, Dibromoacetamide
IUPAC Name 2,2-dibromoacetamide
SMILES NC(=O)C(Br)Br

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C2H3Br2NO/c3-1(4)2(5)6/h1H,(H2,5,6)
Formula Weight 214.8581
Chemical Name or Material Dibromoacetamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.