missing translation for 'onlineSavingsMsg'
Learn More

Dibenzylamine, MP Biomedicals

Product Code. 11416575 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11416575 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11416575 Supplier MP Biomedicals Supplier No. 0520395905

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 103-49-1
Molecular Formula C14H15N
MDL Number MFCD00004770
Quantity 5 g
Synonym dibenzylamine, benzenemethanamine, n-phenylmethyl, n-benzylbenzylamine, bibenzylamine, dibenzyl-amine, dibenzyl amine, bisbenzylamine, n,n-dibenzylamine, n-benzylaminomethyl benzene, unii-3g0yfx01c6
InChI Key BWLUMTFWVZZZND-UHFFFAOYSA-N
SMILES C(NCC1=CC=CC=C1)C1=CC=CC=C1
IUPAC Name dibenzylamine
Molecular Weight (g/mol) 197.28
PubChem CID 7656
Formula Weight 197.28
Chemical Name or Material Dibenzylamine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.