Learn More
Dibenzo[b,d]thiophene 5-oxide, 95%, BLD Pharm™
Specifications
Specifications
| CAS | 1013-23-6 |
| Molecular Formula | C12H8OS |
| MDL Number | MFCD00046902 |
| Quantity | 5 g |
| SMILES | O=S1C2=CC=CC=C2C3=CC=CC=C31 |
| Molecular Weight (g/mol) | 200.26 |
| Percent Purity | 95% |
| Chemical Name or Material | Dibenzo[b,d]thiophene 5-oxide |