Learn More
Dibenzo[b,d]thiophene 5,5-dioxide, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 1016-05-3 |
| Molecular Formula | C12H8O2S |
| MDL Number | MFCD00004970 |
| Quantity | 100 g |
| SMILES | O=S1(C2=CC=CC=C2C3=CC=CC=C31)=O |
| Molecular Weight (g/mol) | 216.26 |
| Percent Purity | 98% |
| Chemical Name or Material | Dibenzo[b,d]thiophene 5,5-dioxide |