Learn More
Diazinon-oxon 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Product Code. 30312111
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30312111 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30312111 Supplier Dr. Ehrenstorfer Supplier No. DREA12210200AL100
 Restricted Item
Estimated Shipment: 21-09-2026
Add to basket
Add to basket
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Diazoxon
CAS 962-58-3
Analyte or Component Names Diazoxon
Shipping Condition Room Temperature
Molecular Formula C12 H21 N2 O4 P
Concentration 100 μg/mL
Certifications/Compliance ISO 17034
Synonym Phosphoric acid, diethyl 6-methyl-2-(1-methylethyl)-4-pyrimidinyl ester, Phosphoric acid, diethyl 2-isopropyl-6-methyl-4-pyrimidinyl ester (6CI,7CI,8CI), 4-Pyrimidinol, 2-isopropyl-6-methyl-, diethyl phosphate (6CI), Diazinon oxon, Diazoxon, Diethyl 2-isopropyl-6-methyl-4-pyrimidinyl phosphate, O,O-Diethyl O-(2-isopropyl-6-methyl-4-pyrimidinyl) phosphate, O,O-Diethyl-O-(2-isopropyl-4-methylpyrimidyl-6) phosphate, Oxodiazinon
InChI Formula InChI=1S/C12H21N2O4P/c1-6-16-19(15,17-7-2)18-11-8-10(5)13-12(14-11)9(3)4/h8-9H,6-7H2,1-5H3
SMILES CCOP(=O)(OCC)Oc1cc(C)nc(n1)C(C)C
IUPAC Name diethyl (6-methyl-2-propan-2-ylpyrimidin-4-yl) phosphate
Solvent or Matrix Acetonitrile
Molecular Weight (g/mol) 288.28
Quantity 1 mL
Formula Weight 288.1239 g/mol