Learn More
VACUUBRAND™ Diaphragm Valve
Product Code. 10273031
Change view
Click to view available options
Diameter (Metric) Outside:
6 mm
16 mm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Outside Unit Size
10273031 6 mm Each
10120941 16 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10273031 Supplier VACUUBRAND™ Supplier No. 20664010
Estimated Shipment: 17-12-2026
Hand operated diaphragm valves for RE 2.5/RZ 2.5/RE 6/RZ 6
VM series valves are hand-operated diaphragm valves. They are used, e.g., as isolation valves, air-admittance valves or manual-control valves. By rotation of the wheel a flexible PTfE diaphragm is moved and pressed against the stainless steel seating or removed from it, closing and opening the valve, respectively. The gas-contacting parts are made of stainless steel and PTfE offering a good resistance to corrosive gases and vapors.
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric) Outside 6 mm
For Use With RE 2.5/RZ 2.5/RE 6/RZ 6
Quantity 1 Each
Type Diaphragm Valve