Learn More
Epredia™ Diamond Point Marker
Product Code. 12804921 Shop All Epredia Products
Change view
Click to view available options
Product Type:
Diamond-point marker
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
12804921 Diamond-point marker Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12804921 Supplier Epredia™ Supplier No. 750
  • ONLINE EXCLUSIVE PRICE
    68.85 EUR
    52.03 EUR / Pack of 1
    Save 16.82 EUR
    24% Off
Estimated Shipment: 18-12-2026
Write indelibly on glass with diamond point marker.
  • Mounted in aluminum pencil
  • 5 in. (12.7cm)
TRUSTED_SUSTAINABILITY
Specifications
Product Type Diamond-point marker
Length (English) 5 in.
Length (Metric) 12.7cm