Learn More
Karl Hecht X100 Diagnostic slides 3 counting fields, approx.D=14mm
Brand:  Karl Hecht 42421010
X100 Diagnostic slides 3 counting fields, approx.D=14mm
Product Code. 10003661
  • ONLINE EXCLUSIVE PRICE
    103.95 EUR
    78.62 EUR / Pack of 100
    Save 25.33 EUR
    24% Off
 Restricted Item
Estimated Shipment: 30-10-2026