Learn More
MP Biomedicals Diagnostic Kit for Myoglobin (MYO) (MPQuanti™ only)
Handla allt MP Biomedicals ProdukterBeskrivning
- Diagnostic Kit for Myoglobin (MYO) (Whole blood/serum/plamsa) is based on fluorescence immunoassay technology.
- The sample moves through the strip from sample pad to absorbent pad.
- If the specimen contains Myoglobin, it attaches to the fluorescent microspheres-conjugated anti-Myoglobin antibodies.
- Then the complex will be captured by the capture antibodies coated on the nitrocellulose membrane (Test line).
- The concentration of Myoglobin in the sample correlates with the fluorescence signal intensity captured on the T line.
- According to the fluorescence intensity of the test and the standard curve, the concentration of Myoglobin in the sample is calculated by the MPQuanti™ Immuflorescence Analyzer.
- CE marked
- For professional in-vitro diagnostic use
- Working with MPQuanti™ Immunofluorescence Analyzer
- Linear working range: 5 ̴ 200 ng/mL, R≥0.990
- Test results should be interpreted at 15 minutes
- For the diagnosis of myocardial infarction (MI)
Specifikationer
Specifikationer
| Detectable Analytes | Myoglobin |
| Detection Method | Immunofluorescence |
| Sample Type | Human whole blood, Human serum, Human plasma |
| Type | POC, Quantitative, Myoglobin, Cardiac |
| Form | Ready-to-Use (RTU) |
Produkttitel
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.
Hittar du en möjlighet till förbättring?