Learn More
Diacetazotol, MedChemExpress
Product Code. 16468866
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16468866 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16468866 Supplier Medchem Express Supplier No. HYB2187
MedChemExpress Diacetazotol inhibits dioxin-induced ethoxyresorufin-O-deethylase (EROD) activity with IC50 of 75±4 nM. Diacetazotol extracts from patent US20070032458, compound 3.
Diacetazotol inhibits dioxin-induced ethoxyresorufin-O-deethylase (EROD) activity with IC50 of 75±4 nM. Diacetazotol extracts from patent US20070032458, compound 3.
Specifications
Chemical Name or Material Diacetazotol
CAS 83-63-6
Color Red
Purity Grade Notes Research
Molecular Formula C18H19N3O2
Quantity 100 mg
Synonym Diacetotoluide
Solubility Information DMSO : 100 mg/mL (323.25 mM; Need ultrasonic)
SMILES CC(N(C(C)=O)C1=CC=C(/N=N/C2=CC=CC=C2C)C=C1C)=O
Molecular Weight (g/mol) 309.36
Formula Weight 309.36
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation