missing translation for 'onlineSavingsMsg'
Learn More

di-Sodium Tetraborate, Supelco™

Product Code. 15874790
Change view
Click to view available options
Quantity:
250 g
1 kg
Unit Size:
1kg
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15874790 1 kg 1kg
15864790 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15874790 Supplier Supelco Supplier No. 1063061000
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For X-ray fluorescence analysis, anhydrous for analysis

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1330-43-4

Specifications

Melting Point 742°C
Quantity 1 kg
MDL Number MFCD00081185
Synonym di-Sodium tetraborate, Borax, di-Sodium tetraborate
Molecular Weight (g/mol) 201.22
Grade Research
Density 2.37 g/cm3 at 20°C
Chemical Name or Material Di-Sodium Tetraborate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.