missing translation for 'onlineSavingsMsg'
Learn More

di-Sodium Succinate Hexahydrate, Sigma-Aldrich™

Product Code. 15830781
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15830781 500 g 500g
15820781 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15830781 Supplier Sigma Aldrich Supplier No. 8201510500
 Restricted Item
Estimated Shipment: 02-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For synthesis

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6106-21-4

Specifications

Melting Point 120°C (Elimination of water of crystallisation)
Quantity 500 g
MDL Number MFCD00149117
Synonym di-Sodium succinate hexahydrate, Succinic acid disodium salt, di-Sodium succinate hexahydrate
Molecular Weight (g/mol) 270.14
Grade Ungraded
Chemical Name or Material Di-Sodium Succinate Hexahydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.