missing translation for 'onlineSavingsMsg'
Learn More

di-Sodium hydrogen phosphate heptahydrate, Supelco™

Product Code. 32238353
Change view
Click to view available options
Quantity:
25 kg
Unit Size:
25kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32238353 25 kg 25kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 32238353 Supplier Supelco Supplier No. 1065759025
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

For analysis EMSURE™ ACS

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material di-Sodium hydrogen phosphate heptahydrate
Melting Point 48°C (Elimination of water of crystallisation)
CAS 7782-85-6
Quantity 25 kg
Density 1.68 g/mL at 25°C (lit.)
Molecular Formula Na2HPO4 · 7H2O
Synonym Sodium phosphate dibasic heptahydrate, Disodium hydrogen phosphate heptahydrate, Disodium phosphate, Sodium phosphate dibasic heptahydrate, Disodium hydrogen phosphate heptahydrate, Disodium phosphate
Molecular Weight (g/mol) 268.07
Grade Not Available
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.