missing translation for 'onlineSavingsMsg'
Learn More
Learn More
di-Sodium Hydrogen Phosphate Dodecahydrate, Supelco™
Specifications
Specifications
| Quantity | 1 kg |
| Molecular Formula | Na2HPO4 · 12H2O |
| MDL Number | MFCD00149181 |
| Synonym | Sodium phosphate dibasic dodecahydrate, Sodium monohydrogen phosphate, Sodium phosphate dibasic, sec-Sodium phosphate, Disodium hydrogen phosphate dodecahydrate, Disodium phosphate, di-Sodium hydrogen phosphate dodecahydrate, Sodium phosphate dibasic dodecahydrate, sec-Sodium phosphate, Disodium hydrogen phosphate dodecahydrate, Disodium phosphate, di-Sodium hydrogen phosphate dodecahydrate |
| Molecular Weight (g/mol) | 358.14 |
| Grade | Research |
| Density | 1.52 g/cm3 at 20°C |
| Chemical Name or Material | Di-Sodium Hydrogen Phosphate Dodecahydrate |
Safety and Handling
Product Identifier
- di-Sodium hydrogen phosphate dodecahydrate for analysis EMSURE® ISO,Reag. Ph Eur
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?