missing translation for 'onlineSavingsMsg'
Learn More
Learn More
di-Sodium Hydrogen Phosphate Dihydrate, Supelco™
Specifications
Specifications
| Melting Point | 92.5°C (Elimination of water of crystallisation) |
| Quantity | 1 kg |
| Molecular Formula | Na2HPO4 · 2H2O |
| MDL Number | MFCD00149182 |
| Synonym | di-Sodium hydrogen phosphate dihydrate, Sodium monohydrogen phosphate, Sodium phosphate dibasic, sec-Sodium phosphate, Disodium hydrogen phosphate dihydrate, Disodium phosphate, Sodium phosphate dibasic dihydrate, di-Sodium hydrogen phosphate dihydrate, sec-Sodium phosphate, Disodium hydrogen phosphate dihydrate, Disodium phosphate, Sodium phosphate dibasic dihydrate |
| Molecular Weight (g/mol) | 177.99 |
| Grade | Research |
| Density | 2.1 g/cm3 at 20°C |
| Chemical Name or Material | Di-Sodium Hydrogen Phosphate Dihydrate |
Safety and Handling
Product Identifier
- di-Sodium hydrogen phosphate dihydrate for analysis EMSURE®
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?