missing translation for 'onlineSavingsMsg'
Learn More

di-Sodium hydrogen phosphate dihydrate, EMSURE™, MilliporeSigma™

Product Code. 15894870
Change view
Click to view available options
Quantity:
50 kg
Packaging:
Fibre Carton
Unit Size:
50kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15894870 50 kg Fibre Carton 50kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15894870 Supplier Supelco Supplier No. 1065809050
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-quality reagent for classical analytical applications.

TRUSTED_SUSTAINABILITY

Specifications

CAS 10028-24-7
Color White
Physical Form Solid
Quantity 50 kg
Molecular Formula H5Na2O6P
Linear Formula Na2HPO4 · 2H2O
MDL Number MFCD00149182
Synonym disodium hydrogen phosphate dihydrate, disodium phosphate dihydrate, sodium phosphate dibasic dihydrate, di-sodium hydrogen phosphate dihydrate, phosphoric acid, disodium salt, dihydrate, sodium hydrogenphosphate dihydrate, sodium phosphate, dibasic, dihydrate, disodium hydrogenphosphate dihydrate, dinatrii phosphas dihydricus, 2na.hpo4.2h2o
Solubility Information 93g/L (20°C)
InChI Key KDQPSPMLNJTZAL-UHFFFAOYSA-L
SMILES O.O.[Na+].[Na+].OP([O-])([O-])=O
IUPAC Name disodium dihydrate hydrogen phosphate
Molecular Weight (g/mol) 177.99
PubChem CID 6451167
Formula Weight 177.99
Percent Purity ≥99.5%
Grade EMSURE
Packaging Fibre Carton
Density 2.1g/cm3 (20°C); Bulk Density: 850 to 1000 kg/m3
Chemical Name or Material Disodium Hydrogen Phosphate Dihydrate, EMSURE™, For Analysis
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.