Learn More
Di-O-cysteinyl-glycinoyl Curcumin 90%, TRC
Specifications
Specifications
| Chemical Name or Material | Di-O-cysteinyl-glycinoyl Curcumin |
| CAS | 1042019-87-3 |
| Molecular Formula | C31H36N4O10S2 |
| Quantity | 1 mg |
| Synonym | 4-((1E,6E)-7-(4-(2-((R)-2-Amino-3-mercaptopropanamido)acetoxy)-3-methoxyphenyl)-3,5-dioxohepta-1,6-dien-1-yl)-2-methoxyphenyl 2-((S)-2-amino-3-mercaptopropanamido)acetate, 2,2'-[[(1E,6E)-3,5-Dioxo-1,6-heptadiene-1,7-diyl]bis(2-methoxy-4,1-phenylene)] Ester L-Cysteinyl-glycine |
| InChI Formula | InChI=1S/C31H36N4O10S2/c1-42-26-11-18(5-9-24(26)44-28(38)14-34-30(40)22(32)16-46)3-7-20(36)13-21(37)8-4-19-6-10-25(27(12-19)43-2)45-29(39)15-35-31(41)23(33)17-47/h3-12,22-23,46-47H,13-17,32-33H2,1-2H3,(H,34,40)(H,35,41)/b7-3+,8-4+/t22-,23-/m0/s1 |
| SMILES | O=C(CC(/C=C/C1=CC(OC)=C(OC(CNC([C@H](N)CS)=O)=O)C=C1)=O)/C=C/C2=CC=C(OC(CNC([C@@H](N)CS)=O)=O)C(OC)=C2 |
| IUPAC Name | 4-((1E,6E)-7-(4-((D-cysteinylglycyl)oxy)-3-methoxyphenyl)-3,5-dioxohepta-1,6-dien-1-yl)-2-methoxyphenyl L-cysteinylglycinate |
| Molecular Weight (g/mol) | 688.77 |
| Formula Weight | 688.19 |
Product Title
¿Detecta una oportunidad de mejora?