missing translation for 'onlineSavingsMsg'
Learn More

di-Ammonium Oxalate Monohydrate, Supelco™

Product Code. 15894040
Change view
Click to view available options
Quantity:
1 kg
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15894040 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15894040 Supplier Supelco Supplier No. 1011901000
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

EMPLURA™

TRUSTED_SUSTAINABILITY

Specifications

Melting Point >123°C (Decomposition)
CAS 6009-70-7
Quantity 1 kg
Molecular Formula (NH4)2C2O4 · H2O
MDL Number MFCD00149694
Synonym di-Ammonium oxalate monohydrate, Oxalic acid ammonium salt, di-Ammonium oxalate monohydrate
Molecular Weight (g/mol) 124.10 (Anhydrous Basis)
Grade Research
Density 1.5 g/cm3 at 20°C
Chemical Name or Material Di-Ammonium Oxalate Monohydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.