missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
This is a rabbit polyclonal anti-DHX32 antibody raised against part of the C-terminal chain of human DHX32.
DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD), are putative RNA helicases. They are implicated in a number of cellular processes involving alteration of RNA secondary structure such as translation initiation, nuclear and mitochondrial splicing, and ribosome and spliceosome assembly. Based on their distribution patterns, some members of this DEAD box protein family are believed to be involved in embryogenesis, spermatogenesis, and cellular growth and division. This gene encodes a member of this family. The function of this member has not been determined. Alternative splicing of this gene generates 2 transcript variants, but the full length nature of one of the variants has not been defined.
Specifications
Specifications
| Antigen | DHX32 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunofluorescence, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.17 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | DHX32 |
| Gene Accession No. | Q7L7V1, Q8BZS9 |
| Gene Alias | DDX32, DEAD/H box 32, DEAD/H helicase like protein 1, DEAH box protein 32, DHLP1, DHX32, HuDDX32 |
| Gene Symbols | Dhx32 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?