missing translation for 'onlineSavingsMsg'
Learn More

DHX32 Antibody - BSA Free, Novus Biologicals™

Product Code. 30559594 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30559594 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30559594 Supplier Novus Biologicals Supplier No. NBP348012

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

DHX32 Polyclonal antibody specifically detects DHX32 in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen DHX32
Applications ELISA, Immunohistochemistry, Western Blot, Immunocytochemistry/Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, ELISA, Immunohistochemistry 1:20-1:200, Immunocytochemistry/ Immunofluorescence 1:20-1:200
Formulation PBS, 50% Glycerol, pH (7.3)
Gene Alias DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 32, DEAD/H box 32, DEAD/H helicase-like protein 1, DEAH (Asp-Glu-Ala-His) box polypeptide 32, DEAH box protein 32, DHLP1DDX32DEAD/H helicase-like protein-1, EC 3.6.1, EC 3.6.4.13, FLJ10694, FLJ10889, huDDX32, putative pre-mRNA-splicing factor ATP-dependent RNA helicase DHX32
Host Species Rabbit
Immunogen DEAH(Asp-Glu-Ala-His) box polypeptide 32. (Uniprot# Q7L7V1)
Purification Method Immunogen affinity purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55760
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.