Learn More
(DHQ)2PHAL, TRC
Product Code. 30180113
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
Unit Size:
100mg
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30180113 100 mg 100mg
30180049 500 mg 500mg
30179991 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30180113 Supplier Toronto Research Chemicals Supplier No. TRCD299920100MG
 Restricted Item
Estimated Shipment: 11-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 140924-50-1
Molecular Formula C48H54N6O4
Molecular Weight (g/mol) 778.98
Synonym 1,4-Bis(9-O-dihydroquininyl)phthalazine, 1,4-Bis(9-O-dihydroquinyl)phthalazine, 1,4-Bis(dihydroquinine)phthalazine, (8alpha,9R)-(8' 'alpha,9' 'R)-9,9' '-[1,4-Phthalazinediylbis(oxy)]bis[10,11-dihydro-6'-methoxycinchonan];
SMILES CC[C@H]1CN2CC[C@H]1C[C@H]2[C@H](Oc3nnc(O[C@@H]([C@@H]4C[C@@H]5CCN4C[C@@H]5CC)c6ccnc7ccc(OC)cc67)c8ccccc38)c9ccnc%10ccc(OC)cc9%10
Specifications
Quantity 100 mg
InChI Formula InChI=1S/C48H54N6O4/c1-5-29-27-53-21-17-31(29)23-43(53)45(35-15-19-49-41-13-11-33(55-3)25-39(35)41)57-47-37-9-7-8-10-38(37)48(52-51-47)58-46(44-24-32-18-22-54(44)28-30(32)6-2)36-16-20-50-42-14-12-34(56-4)26-40(36)42/h7-16,19-20,25-26,29-32,43-46H,5-6,17-18,21-24,27-28H2,1-4H3/t29-,30+,31-,32+,43-,44-,45-,46-/m1/s1
Formula Weight 778.4207
Chemical Name or Material (DHQ)2PHAL