missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DHDH Polyclonal Antibody
GREENER_CHOICE

Product Code. 17151364 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17151364 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17151364 Supplier Invitrogen™ Supplier No. PA5113505

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

DHDH Polyclonal Antibody for Western Blot, IHC (P), ELISA

This gene encodes an enzyme that belongs to the family of dihydrodiol dehydrogenases, which exist in multiple forms in mammalian tissues and are involved in the metabolism of xenobiotics and sugars. These enzymes catalyze the NADP1-linked oxidation of transdihydrodiols of aromatic hydrocarbons to corresponding catechols. This enzyme is a dimeric dihydrodiol dehydrogenase, and it differs from monomeric dihydrodiol dehydrogenases in its high substrate specificity for trans-dihydrodiols of aromatic hydrocarbons in the oxidative direction.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DHDH
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.29 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene DHDH
Gene Accession No. Q9DBB8, Q9UQ10
Gene Alias 1300018L09Rik; 2DD; 3-deoxyglucosone reductase; AA591799; CAN2DD; ch211-203b17.3; DDH; DHDH; dhdh.2; dhdh-like.1.L; dihydrodiol dehydrogenase; dihydrodiol dehydrogenase (dimeric); dihydrodiol dehydrogenase, tandem duplicate 2; dimeric dihydrodiol dehydrogenase; D-xylose 1-dehydrogenase; D-xylose-NADP dehydrogenase; Hum2DD; putative LOC617564 protein; SUS2DD; Trans-1,2-dihydrobenzene-1,2-diol dehydrogenase; trans-1,2-dihydrobenzene-1,2-diol dehydrogenase L homeolog; XELAEV_18036112mg; Xld1; Xld-1; xylose dehydrogenase 1; zgc:101723
Gene Symbols DHDH
Host Species Rabbit
Immunogen Recombinant Human Trans-1,2-dihydrobenzene-1,2-diol dehydrogenase protein (1-334AA).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27294, 71755
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.