Learn More
R&D Systems™ DGCR8 knockout Mouse embryonic stem cells
Product Code. 18614030
Change view
Click to view available options
Quantity:
2 mL
Unit Size:
2mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18614030 2 mL 2mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18614030 Supplier R&D Systems™ Supplier No. NBA119349
  • ONLINE EXCLUSIVE PRICE
    694.00 EUR
    656.25 EUR / 2mL
    Save 37.75 EUR
    5% Off
 Restricted Item
Estimated Shipment: 13-11-2026
A Novus product, part of R&D Systems.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store in gas phase of liquid nitrogen.
Species Mouse
For Use With (Application) Flow Cytometry, Immunocytochemistry/ Immunofluorescence, In vitro assay, In vivo assay
Product Line Novus
Quantity 2 mL