Learn More
Dexrazoxane, MedChemExpress
Product Code. 16428538
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16428538 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16428538 Supplier Medchem Express Supplier No. HYB0581
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
MedChemExpress Dexrazoxane (ICRF-187) is a cardioprotective agent.
Dexrazoxane (ICRF-187) is a cardioprotective agent.
Specifications
Chemical Name or Material Dexrazoxane
CAS 24584-09-6
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C11H16N4O4
Quantity 10 mg
Synonym ICRF-187 ADR-529 NSC-169780
Solubility Information DMSO : 25 mg/mL (93.19 mM; Need ultrasonic)
SMILES C[C@H](N(C1)CC(NC1=O)=O)CN(C2)CC(NC2=O)=O
Molecular Weight (g/mol) 268.27
Formula Weight 268.27
Percent Purity 98.01%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death