Learn More
Dexamethasone EP Impurity D (Mometasone Furoate EP Impurity L), Mikromol™
Description
Dexamethasone EP Impurity D (Mometasone Furoate EP Impurity L)
Specifications
Specifications
| Chemical Name or Material | 9beta,11beta-Epoxy-17,21-dihydroxy-16alpha-methylpregna-1,4-diene-3,20-dione |
| CAS | 24916-90-3 |
| Analyte or Component Names | 9beta,11beta-Epoxy-17,21-dihydroxy-16alpha-methylpregna-1,4-diene-3,20-dione |
| Shipping Condition | Room Temperature |
| Molecular Formula | C22 H28 O5 |
| Certifications/Compliance | ISO 17025 |
| Synonym | 9β,11β-Epoxy-17,21-dihydroxy-16α-methylpregna-1,4-diene-3,20-dione, Dexamethasone Imp. D (EP), 17,21-Dihydroxy-16alpha-methyl-9beta,11beta-epoxypregna-1,4-diene-3,20-dione, 9beta,11beta-Epoxy-17,21-dihydroxy-16alpha-methylpregna-1,4-diene-3,20-dione, Dexamethasone 9,11-Epoxide, Dexamethasone Impurity D, Mometasone Furoate Impurity L |
| InChI Formula | InChI=1S/C22H28O5/c1-12-8-16-15-5-4-13-9-14(24)6-7-19(13,2)22(15)18(27-22)10-20(16,3)21(12,26)17(25)11-23/h6-7,9,12,15-16,18,23,26H,4-5,8,10-11H2,1-3H3/t12-,15+,16+,18+,19+,20+,21+,22-/m1/s1 |
| SMILES | C[C@@H]1C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]35O[C@H]5C[C@]2(C)[C@@]1(O)C(=O)CO |
| IUPAC Name | 17,21-Dihydroxy-16α-methyl-9β,11β-epoxypregna-1,4-diene-3,20-dione |
| Show More |