Learn More
Dexamethasone Acetate, LoGiCal
Specifications
Specifications
| Chemical Name or Material | Dexamethasone Acetate |
| CAS | 1177-87-3 |
| Analyte or Component Names | Dexamethasone Acetate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C24 H31 F O6 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Betamethasone Acetate Imp. B (EP), Dexamethasone Imp. G (EP), Betamethasone Acetate Impurity B, Dexamethasone Impurity G, Dexamethasone Isonicotinate Impurity B |
| InChI Formula | InChI=1S/C24H31FO6/c1-13-9-18-17-6-5-15-10-16(27)7-8-21(15,3)23(17,25)19(28)11-22(18,4)24(13,30)20(29)12-31-14(2)26/h7-8,10,13,17-19,28,30H,5-6,9,11-12H2,1-4H3/t13-,17+,18+,19+,21+,22+,23+,24+/m1/s1 |
| Solution Type | Neat |
| SMILES | C[C@@H]1C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]2(C)[C@@]1(O)C(=O)COC(=O)C |
| Show More |