missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Dexamethasone Sodium Phosphate is used in the treatment of emetic effects of cancer therapy and epicondylitis.
Specifications
Specifications
| CAS | 2392-39-4 |
| Physical Form | Powder |
| Quantity | 5 g |
| Molecular Formula | C22H28FNa2O8P |
| Synonym | dexamethasone sodium phosphate, dexamethasone 21-phosphate disodium salt, dalalone, dexadreson, dexagro, megacort, soldesam, spersadox, decdan, solu-decadron |
| Solubility Information | Soluble in water (200mg/4mL, clear, faint yellow solution). |
| InChI Key | PLCQGRYPOISRTQ-FCJDYXGNSA-L |
| SMILES | CC1CC2C3CCC4=CC(=O)C=CC4(C3(C(CC2(C1(C(=O)COP(=O)([O-])[O-])O)C)O)F)C.[Na+].[Na+] |
| IUPAC Name | disodium;[2-[(8S,9R,10S,11S,13S,14S,16R,17R)-9-fluoro-11,17-dihydroxy-10,13,16-trimethyl-3-oxo-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-17-yl]-2-oxoethyl] phosphate |
| Molecular Weight (g/mol) | 516.41 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?