Learn More
KGW Isotherm™ Dewar Insulated Carrying Flasks
Product Code. 10312861
Click to view available options
Capacity (Metric):
1 L
2 L
3 L
Cap Lid Type:
Blue-coated Metal Cover
Stainless Steel Cover
Unit Size:
Each
Product Code. 10312861
Brand: KGW Isotherm™ 1212
Only null left
Add to basket
Add to basket
For the storage and transport of temperature-sensitive goods such as dry ice. KGW Isotherm™ Dewar Insulated Carrying Flasks feature a robust design made from borosilicate glass 3.3.
  • Cooling of small samples
  • Silver-plated and evacuated
  • Flexible applications in the laboratory
  • Includes handle made from aluminum
  • Includes insulated lid, held by two spring clips
  • Available with either blue-coated metal cover or stainless steel cover
Specifications
Cap Lid Type Blue-coated Metal Cover
Diameter (Metric) Outer 160mm
For Use With (Application) Cooling of small samples
Height (Metric) Inner 170mm
Height (Metric) 245mm
Includes Insulated lid, held by two spring clips, handle
Capacity (Metric) 2 L
Diameter (Metric) Inner 138 mm
Material Borosilicate Glass