missing translation for 'onlineSavingsMsg'
Learn More

Devarda's Alloy, For analysis, EMSURE™, MilliporeSigma™

Product Code. 15864550
Change view
Click to view available options
Quantity:
1 kg
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15864550 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15864550 Supplier Supelco Supplier No. 1053411000
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 8049-11-4
Appearance Gray solid
Quantity 1 kg
Odor Odorless
Packaging Plastic bottle
Form Solid
Molecular Formula Al9Cu10Zn
Synonym devarda's alloy, devarda's alloy, granular, al.cu.zn, devarda's alloy, p.a., alumane copper zinc, devarda's alloy, puriss. p.a., devarda's alloy, saj special grade, devarda's alloy, powder,-100 mesh, devarda's alloy, filings, +100 mesh, devarda inverted exclamation mark s alloy
InChI Key MPKKOQSNGWAMJD-UHFFFAOYSA-N
SMILES [Al].[Cu].[Zn]
IUPAC Name aluminum;copper;zinc
Molecular Weight (g/mol) 943.67
PubChem CID 16211762
Formula Weight 5.79. (molar mass)
Chemical Name or Material Devarda's Alloy
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.