Learn More
Detritylation Reagent, Solstice
Product Code. 16825701
Change view
Click to view available options
4X2.5L GLASS BOTTLE
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
16825701 4X2.5L GLASS BOTTLE
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16825701 Supplier Burdick & Jackson BioSyn Supplier No. BR6054x2.5L
  • ONLINE EXCLUSIVE PRICE
    1675.00 EUR
    411.25 EUR / Pack of 4
    Save 1263.75 EUR
    75% Off
 Restricted Item
Request Bulk or Custom Format
Currently Unavailable: We have no information when this product will be available.
View alternative products
BioSyn™, suitable for use with DNA/RNA synthesis instruments, 3%Trichloroacetic Acid in Dichloromethane (withv)
Solstice BioSyn™ reagents and solvents for DNA/RNA synthesis
  • Solstice BioSyn™ Solvents and Reagents are specifically processed and purified to exacting specifications, ensuring outstanding synthesis efficiency
  • ideal for oligonucleotide synthesis regardless of the application
  • All solvents and reagents are filtered (0.2-micron for oxidation reagents and 0.1-micron for all other reagents) to minimize particulate contamination
TRUSTED_SUSTAINABILITY
Specifications
CAS 75-09-2, 76-03-9
CAS Max % 97.00%, 3.00%
Quantity 4 x 2.5 L
Packaging Glass Bottle
Physical Form Liquid, clear
Purity Grade Notes Deblocking reagents
Boiling Point 40°C