Learn More
Detomidine Hydrochloride, TRC
Product Code. 30135842
Change view
Click to view available options
Quantity:
10 mg
100 mg
1 g
Unit Size:
100mg
10mg
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135842 1 g 1g
18022040 100 mg 100mg
18053640 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30135842 Supplier Toronto Research Chemicals Supplier No. TRCD2979751G
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 90038-01-0
Molecular Formula C12 H14 N2 . Cl H
Molecular Weight (g/mol) 222.71
Synonym Detomidine hydrochloride, 4-(2,3-Dimethylbenzyl)-1H-imidazole hydrochloride, 1H-Imidazole, 4-[(2,3-dimethylphenyl)methyl]-, monohydrochloride, Detomidine hydrochloride, Domosedan, Dormosedan, MPV 253AII
IUPAC Name 4-[(2,3-dimethylphenyl)methyl]-1H-imidazole;hydrochloride
SMILES Cl.Cc1cccc(Cc2c[nH]cn2)c1C
Specifications
Quantity 1 g
InChI Formula InChI=1S/C12H14N2.ClH/c1-9-4-3-5-11(10(9)2)6-12-7-13-8-14-12;/h3-5,7-8H,6H2,1-2H3,(H,13,14);1H
Formula Weight 222.0924
Chemical Name or Material Detomidine hydrochloride