Learn More
Despyridyl Piroxicam (Piroxicam Impurity C), TRC
Specifications
Specifications
| Chemical Name or Material | 4-Hydroxy-2-methyl-2H-1,2-benzothiazine-3-carboxamide 1,1-Dioxide |
| CAS | 24683-25-8 |
| Molecular Formula | C10 H10 N2 O4 S |
| Quantity | 50 mg |
| Synonym | 4-Hydroxy-2-methyl-2H-1,2-benzothiazine-3-carboxamide 1,1-Dioxide, Piroxicam Imp. C (EP) |
| InChI Formula | InChI=1S/C10H10N2O4S/c1-12-8(10(11)14)9(13)6-4-2-3-5-7(6)17(12,15)16/h2-5,13H,1H3,(H2,11,14) |
| SMILES | CN1C(=C(O)c2ccccc2S1(=O)=O)C(=O)N |
| IUPAC Name | 4-hydroxy-2-methyl-1,1-dioxo-1λ^{6},2-benzothiazine-3-carboxamide |
| Molecular Weight (g/mol) | 254.26 |
| Formula Weight | 254.0361 |