Learn More
Desphenyl Chloridazon, TRC
Product Code. 17994325
Change view
Click to view available options
Quantity:
5 mg
10 mg
100 mg
Unit Size:
100mg
10mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17994325 100 mg 100mg
17984495 5 mg 5mg
17994075 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17994325 Supplier Toronto Research Chemicals Supplier No. TRCD297000100MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 6339-19-1
Molecular Formula C4 H4 Cl N3 O
Molecular Weight (g/mol) 145.55
Synonym 3-Pyridazinol, 5-amino-4-chloro- (6CI,7CI), 5-Amino-4-chloro-3(2H)-pyridazinone, 4-Amino-5-chloropyridazin-6(1H)-one, 5-Amino-4-chloro-3(2H)-pyridazone, 5-Amino-4-chloro-3-pyridazinone, NSC 38293
IUPAC Name 4-amino-5-chloro-1H-pyridazin-6-one
SMILES NC1=C(Cl)C(=O)NN=C1
Specifications
Quantity 100 mg
InChI Formula InChI=1S/C4H4ClN3O/c5-3-2(6)1-7-8-4(3)9/h1H,(H3,6,8,9)
Formula Weight 145.0043
Percent Purity >95%
Chemical Name or Material Chloridazon-desphenyl
Grade HPLC