missing translation for 'onlineSavingsMsg'
Learn More

Desphenyl Chloridazon, TRC

Product Code. 17984495
Change view
Click to view available options
Quantity:
5 mg
10 mg
100 mg
Unit Size:
100mg
10mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17984495 5 mg 5mg
17994325 100 mg 100mg
17994075 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17984495 Supplier Toronto Research Chemicals Supplier No. TRCD2970005MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6339-19-1
Molecular Formula C4 H4 Cl N3 O
Molecular Weight (g/mol) 145.55
Synonym 3-Pyridazinol, 5-amino-4-chloro- (6CI,7CI), 5-Amino-4-chloro-3(2H)-pyridazinone, 4-Amino-5-chloropyridazin-6(1H)-one, 5-Amino-4-chloro-3(2H)-pyridazone, 5-Amino-4-chloro-3-pyridazinone, NSC 38293
IUPAC Name 4-amino-5-chloro-1H-pyridazin-6-one
SMILES NC1=C(Cl)C(=O)NN=C1

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C4H4ClN3O/c5-3-2(6)1-7-8-4(3)9/h1H,(H3,6,8,9)
Formula Weight 145.0043
Percent Purity >95%
Chemical Name or Material Chloridazon-desphenyl
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.