Learn More
Desmethylsibutramine Hydrochloride, LoGiCal
Product Code. 30459802
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30459802 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30459802 Supplier LoGiCal Supplier No. LGCFOR0991.02
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Desmethylsibutramine Hydrochloride
CAS 84467-94-7
Analyte or Component Names Desmethylsibutramine Hydrochloride
Shipping Condition Room Temperature
Molecular Formula C16 H24 Cl N . Cl H
Certifications/Compliance ISO 9001
Synonym (1RS)-1-[1-(4-Chlorophenyl)cyclobutyl]-N,3-dimethylbutan-1-amine Hydrochloride, Sibutramine Hydrochloride Monohydrate Imp. D (Pharmeuropa) as Hydrochloride, N-Monodesmethylsibutramine Hydrochloride, Desmethylsibutramine Hydrochloride, Sibutramine Related Compound D as Hydrochloride, Sibutramine Hydrochloride Monohydrate Impurity D, Sibutramine Impurity D
InChI Formula InChI=1S/C16H24ClN.ClH/c1-12(2)11-15(18-3)16(9-4-10-16)13-5-7-14(17)8-6-13;/h5-8,12,15,18H,4,9-11H2,1-3H3;1H
Solution Type Neat
SMILES Cl.CNC(CC(C)C)C1(CCC1)c2ccc(Cl)cc2
IUPAC Name 1-[1-(4-chlorophenyl)cyclobutyl]-N,3-dimethylbutan-1-amine;hydrochloride
Industry Type Clinical Testing
Molecular Weight (g/mol) 302.28
Quantity 10 mg
Formula Weight 301.1364 g/mol